Assay Design



Gene Transcripts

Gene Symbol
ARF11
Entrez Gene ID
819264
Gene Name
auxin response factor 11
Gene Aliases
AT2G46530, F11C10.34, auxin response factor 11
Chromosome Location
Chr.2: 19104632 - 19108486 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130218.5 NP_182176.2 1 - 2 392 67
NM_201972.1 NP_973701.1 1 - 2 341 67
GenBank mRNA AK229924.1 - 1 - 2 203 67

View Full Product Details