Assay Design



Gene Transcripts

Gene Symbol
PIL1
Entrez Gene ID
819311
Gene Name
phytochrome interacting factor 3-like 1
Gene Aliases
AT2G46970, F14M4.20, phytochrome interacting factor 3-like 1
Chromosome Location
Chr.2: 19295431 - 19297858 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337226.1 NP_001323544.1 2 - 3 748 75
NM_001337227.1 NP_001323543.1 3 - 4 673 75
NM_130265.4 NP_182220.2 3 - 4 903 75
GenBank mRNA AY954840.1 - 3 - 4 723 75

View Full Product Details