Assay Design
Gene Transcripts
- Gene Symbol
- NF-YB5
- Entrez Gene ID
-
819393
- Gene Name
- nuclear factor Y, subunit B5
- Gene Aliases
-
''nuclear factor Y, AT2G47810, F17A22.20, nuclear factor Y, subunit B5, subunit B5''
- Chromosome Location
-
Chr.2: 19582658 - 19583618 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130348.3 |
NP_182302.1 |
1 - 1 |
727 |
156 |
GenBank mRNA |
BT003968.1 |
- |
1 - 1 |
727 |
156 |
View Full Product Details