Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130365.4 | NP_566119.1 | 4 - 5 | 637 | 134 |
GenBank mRNA | AJ242965.1 | - | 4 - 5 | 511 | 134 |
AY063915.1 | - | 4 - 5 | 536 | 134 | |
AY091270.1 | - | 4 - 5 | 421 | 134 |