Assay Design



Gene Transcripts

Gene Symbol
SCC3
Entrez Gene ID
819410
Gene Name
sister-chromatid cohesion protein 3
Gene Aliases
AT2G47980, ATSCC3, SISTER-CHROMATID COHESION PROTEIN 3, T9J23.3, sister-chromatid cohesion protein 3
Chromosome Location
Chr.2: 19631207 - 19636978 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130365.4 NP_566119.1 5 - 6 793 72
GenBank mRNA AJ242965.1 - 5 - 6 667 72
AY063915.1 - 5 - 6 692 72
AY091270.1 - 5 - 6 577 72

View Full Product Details