Assay Design



Gene Transcripts

Gene Symbol
RNL
Entrez Gene ID
837306
Gene Name
RNAligase
Gene Aliases
ARABIDOPSIS THALIANA RNA LIGASE, AT1G07910, ATRNL, AtRLG1, RNAligase, T6D22.1, T6D22_1, TRNA LIGASE
Chromosome Location
Chr.1: 2445437 - 2454645 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331741.1 NP_001318950.1 9 - 10 1431 110
NM_100665.3 NP_172269.2 9 - 10 1309 110
GenBank mRNA AK230326.1 - 9 - 10 1276 110

View Full Product Details