Assay Design



Gene Transcripts

Gene Symbol
LIG1
Entrez Gene ID
837333
Gene Name
DNA ligase 1
Gene Aliases
AT1G08130, ATLIG1, DNA ligase 1, T6D22.23
Chromosome Location
Chr.1: 2542663 - 2547946 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100689.5 NP_172293.2 11 - 12 2079 139
GenBank mRNA AK117238.1 - 11 - 12 2087 139
BT005964.1 - 11 - 12 1948 139

View Full Product Details