Assay Design



Gene Transcripts

Gene Symbol
CPNIFS
Entrez Gene ID
837370
Gene Name
chloroplastic NIFS-like cysteine desulfurase
Gene Aliases
AT1G08490, ATCPNIFS, ATNFS2, ATSUFS, CYSTEINE DESULFURASE, SUFS, T27G7.17, T27G7_17, chloroplastic NIFS-like cysteine desulfurase
Chromosome Location
Chr.1: 2685659 - 2688658 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100722.5 NP_172325.2 2 - 3 604 97
GenBank mRNA AF419347.1 - 2 - 3 493 97
AY078068.1 - 2 - 3 493 97
AY094425.1 - 2 - 3 535 97
AY149950.1 - 2 - 3 493 97

View Full Product Details