Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_100722.5 | NP_172325.2 | 6 - 7 | 1071 | 96 |
GenBank mRNA | AF419347.1 | - | 6 - 7 | 960 | 96 |
AY078068.1 | - | 6 - 7 | 960 | 96 | |
AY094425.1 | - | 6 - 7 | 1002 | 96 | |
AY149950.1 | - | 6 - 7 | 960 | 96 |