Assay Design



Gene Transcripts

Gene Symbol
SNL6
Entrez Gene ID
837584
Gene Name
paired amphipathic helix Sin3-like protein
Gene Aliases
AT1G10450, SIN3-like 6, T10O24.5, T10O24_5
Chromosome Location
Chr.1: 3431764 - 3438370 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331906.1 NP_001321764.1 2 - 3 434 78
NM_001331907.1 NP_001321765.1 1 - 2 221 78
NM_100919.5 NP_172515.2 2 - 3 434 78

View Full Product Details