Assay Design



Gene Transcripts

Gene Symbol
SEX1
Entrez Gene ID
837619
Gene Name
Pyruvate phosphate dikinase, PEP/pyruvate binding domain-containing protein
Gene Aliases
AT1G10760, GWD, GWD1, SOP, SOP1, STARCH EXCESS 1, T16B5.10, T16B5_10
Chromosome Location
Chr.1: 3580830 - 3590928 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331926.1 NP_001318975.1 32 - 33 4340 80
NM_001331927.1 NP_001320611.1 31 - 32 4189 80
NM_100952.4 NP_563877.1 33 - 34 4174 80
GenBank mRNA AF312027.1 - 30 - 31 3880 80

View Full Product Details