Assay Design



Gene Transcripts

Gene Symbol
RECQ4A
Entrez Gene ID
837636
Gene Name
DNA helicase (RECQl4A)
Gene Aliases
AT1G10930, ATRECQ4A, ATSGS1, T19D16.15, T19D16_15
Chromosome Location
Chr.1: 3647876 - 3655685 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100968.3 NP_172562.2 4 - 5 547 101
GenBank mRNA AY120761.1 - 4 - 5 445 101
BT010133.1 - 3 - 4 241 101

View Full Product Details