Assay Design



Gene Transcripts

Gene Symbol
CSN6A
Entrez Gene ID
835727
Gene Name
COP9 signalosome subunit 6A
Gene Aliases
AT5G56280, COP9 signalosome subunit 6A, MXK23.2, MXK23_2
Chromosome Location
Chr.5: 22783384 - 22785605 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125011.4 NP_568839.1 7 - 8 887 115
GenBank mRNA AY048692.1 - 7 - 8 853 115
BT025877.1 - 7 - 8 812 115

View Full Product Details