Assay Design



Gene Transcripts

Gene Symbol
EXPA14
Entrez Gene ID
835731
Gene Name
expansin A14
Gene Aliases
AT5G56320, ATEXP14, ATEXPA14, ATHEXP ALPHA 1.5, EXP14, EXPANSIN 14, MCD7.4, MCD7_4, expansin A14
Chromosome Location
Chr.5: 22808641 - 22810747 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345194.1 NP_001332569.1 2 - 3 665 110
NM_001345195.1 NP_001332570.1 3 - 4 556 110
NM_125015.2 NP_200443.1 3 - 4 556 110
GenBank mRNA AK230274.1 - 3 - 4 556 110

View Full Product Details