Assay Design



Gene Transcripts

Gene Symbol
SYNC1
Entrez Gene ID
835769
Gene Name
Class II aminoacyl-tRNA and biotin synthetases superfamily protein
Gene Aliases
AT5G56680, EMB2755, EMBRYO DEFECTIVE 2755, MIK19.13, MIK19_13, SYNC1 ARATH
Chromosome Location
Chr.5: 22936534 - 22939060 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125051.4 NP_200479.1 1 - 2 606 135
GenBank mRNA AF170909.1 - 1 - 2 495 135
BT030358.1 - 1 - 2 495 135

View Full Product Details