Assay Design
Gene Transcripts
- Gene Symbol
- ATLIG4
- Entrez Gene ID
-
835822
- Gene Name
- DNA ligase IV
- Gene Aliases
-
AT5G57160, DNA LIGASE IV, LIG4
- Chromosome Location
-
Chr.5: 23154965 - 23161826 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125098.4 |
NP_568851.2 |
6 - 7 |
954 |
117 |
GenBank mRNA |
AF233527.1 |
- |
6 - 7 |
874 |
117 |
View Full Product Details