Assay Design



Gene Transcripts

Gene Symbol
ORP4C
Entrez Gene ID
835830
Gene Name
OSBP(oxysterol binding protein)-related protein 4C
Gene Aliases
AT5G57240, MJB24.5, MJB24_5, OSBP(oxysterol binding protein)-related protein 4C
Chromosome Location
Chr.5: 23192872 - 23195476 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037011.2 NP_001032088.1 3 - 4 559 69
NM_001345250.1 NP_001329986.1 2 - 3 966 69
NM_001345251.1 NP_001329984.1 3 - 4 757 69
NM_001345252.1 NP_001329985.1 3 - 4 757 69
NM_125106.3 NP_200534.1 2 - 3 966 69
GenBank mRNA AK117264.1 - 2 - 3 517 69

View Full Product Details