Assay Design
Gene Transcripts
- Gene Symbol
- XRCC3
- Entrez Gene ID
-
835850
- Gene Name
- homolog of X-ray repair cross complementing 3 (XRCC3)
- Gene Aliases
-
ARABIDOPSIS THALIANA HOMOLOG OF X-RAY REPAIR CROSS COMPLEMENTING 3 (XRCC3), AT5G57450, ATXRCC3, MUA2.3, MUA2_3, homolog of X-ray repair cross complementing 3, homolog of X-ray repair cross complementing 3 (XRCC3)
- Chromosome Location
-
Chr.5: 23273067 - 23274976 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125127.3 |
NP_200554.1 |
1 - 1 |
1116 |
79 |
GenBank mRNA |
AB073492.1 |
- |
1 - 1 |
1116 |
79 |
View Full Product Details