Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001345268.1 | NP_001330674.1 | 7 - 8 | 1216 | 128 |
NM_001345269.1 | NP_001330673.1 | 6 - 7 | 781 | 128 | |
NM_125144.4 | NP_568860.1 | 7 - 8 | 1290 | 128 | |
GenBank mRNA | AY034100.1 | - | 7 - 8 | 944 | 128 |