Assay Design
Gene Transcripts
- Gene Symbol
- TET15
- Entrez Gene ID
-
835890
- Gene Name
- tetraspanin15
- Gene Aliases
-
AT5G57810, MTI20.4, MTI20_4, tetraspanin15
- Chromosome Location
-
Chr.5: 23424268 - 23425647 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125165.3 |
NP_568866.1 |
1 - 2 |
678 |
97 |
GenBank mRNA |
DQ447087.1 |
- |
1 - 2 |
620 |
97 |
View Full Product Details