Assay Design
Gene Transcripts
- Gene Symbol
- UBP23
- Entrez Gene ID
-
835910
- Gene Name
- ubiquitin-specific protease 23
- Gene Aliases
-
AT5G57990, MTI20.25, MTI20_25, ubiquitin-specific protease 23
- Chromosome Location
-
Chr.5: 23469924 - 23474172 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125184.3 |
NP_568873.1 |
5 - 6 |
1213 |
95 |
View Full Product Details