Assay Design



Gene Transcripts

Gene Symbol
CPL4
Entrez Gene ID
5008312
Gene Name
C-terminal domain phosphatase-like 4
Gene Aliases
AT5G58003, C-terminal domain phosphatase-like 4
Chromosome Location
Chr.5: 23479781 - 23482136 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085295.2 NP_001078764.1 3 - 4 302 95
GenBank mRNA DQ503426.1 - 2 - 3 171 95

View Full Product Details