Assay Design



Gene Transcripts

Gene Symbol
ATYKT62
Entrez Gene ID
835930
Gene Name
Synaptobrevin family protein
Gene Aliases
AT5G58180, MCK7.5, MCK7_5, YKT62
Chromosome Location
Chr.5: 23543652 - 23545760 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345292.1 NP_001318826.1 2 - 3 296 64
NM_125203.2 NP_200626.1 1 - 2 192 64

View Full Product Details