Assay Design
Gene Transcripts
- Gene Symbol
- EXO70B1
- Entrez Gene ID
-
835956
- Gene Name
- exocyst subunit exo70 family protein B1
- Gene Aliases
-
AT5G58430, ATEXO70B1, MQJ2.2, MQJ2_2, exocyst subunit exo70 family protein B1
- Chromosome Location
-
Chr.5: 23621152 - 23623543 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125229.4 |
NP_200651.1 |
1 - 1 |
2228 |
159 |
View Full Product Details