Assay Design
Gene Transcripts
- Gene Symbol
- PLC1
- Entrez Gene ID
-
835981
- Gene Name
- phospholipase C1
- Gene Aliases
-
ARABIDOPSIS THALIANA PHOSPHOLIPASE C, AT5G58670, ATPLC, ATPLC1, MZN1.12, MZN1_12, PHOSPHOLIPASE C, phospholipase C 1, phospholipase C1
- Chromosome Location
-
Chr.5: 23704231 - 23707003 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125254.2 |
NP_568881.1 |
6 - 7 |
1551 |
100 |
GenBank mRNA |
D38544.1 |
- |
6 - 7 |
1348 |
100 |
View Full Product Details