Assay Design



Gene Transcripts

Gene Symbol
MBD7
Entrez Gene ID
836101
Gene Name
methyl-CPG-binding domain 7
Gene Aliases
ARABIDOPSIS THALIANA METHYL-CPG-BINDING DOMAIN 7, AT5G59800, ATMBD7, MMN10.2, MMN10_2, methyl-CPG-binding domain 7
Chromosome Location
Chr.5: 24094359 - 24096532 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125372.4 NP_200788.1 2 - 3 421 77
GenBank mRNA BT014975.1 - 2 - 3 340 77
BT015848.1 - 2 - 3 311 77

View Full Product Details