Assay Design



Gene Transcripts

Gene Symbol
NRPC1
Entrez Gene ID
836126
Gene Name
nuclear RNA polymerase C1
Gene Aliases
AT5G60040, MGO3.13, MGO3_13, nuclear RNA polymerase C1
Chromosome Location
Chr.5: 24173259 - 24183641 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203644.1 NP_001190573.1 2 - 3 285 81
NM_125397.3 NP_200812.2 2 - 3 181 81

View Full Product Details