Assay Design



Gene Transcripts

Gene Symbol
AGL8
Entrez Gene ID
836212
Gene Name
AGAMOUS-like 8
Gene Aliases
AGAMOUS-like 8, AT5G60910, FRUITFULL, FUL, MSL3.30, MSL3_30
Chromosome Location
Chr.5: 24502434 - 24506269 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085307.1 NP_001078776.1 7 - 8 447 76
NM_125484.4 NP_568929.1 7 - 8 872 76
GenBank mRNA AF386929.1 - 7 - 8 675 76
AY072463.1 - 7 - 8 616 76
U33473.1 - 7 - 8 716 76

View Full Product Details