Assay Design



Gene Transcripts

Gene Symbol
ABCA11
Entrez Gene ID
836291
Gene Name
ABC2 homolog 15
Gene Aliases
ABC2 homolog 15, ARABIDOPSIS THALIANA ABC2 HOMOLOG 15, AT5G61690, ATATH15, ATH15, ATP-binding cassette A11, K11J9.22, K11J9_22
Chromosome Location
Chr.5: 24789237 - 24793528 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345487.1 NP_001318858.1 11 - 12 2409 94
NM_001345488.1 NP_001332171.1 11 - 12 2412 94

View Full Product Details