Assay Design



Gene Transcripts

Gene Symbol
ABCA12
Entrez Gene ID
836292
Gene Name
ABC2 homolog 16
Gene Aliases
ABC2 homolog 16, ARABIDOPSIS THALIANA ABC2 HOMOLOG 16, AT5G61700, ATATH16, ATH16, ATP-binding cassette A12, K11J9.11, K11J9_11
Chromosome Location
Chr.5: 24793779 - 24798745 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345489.1 NP_001318859.1 12 - 13 2128 114
NM_001345490.1 NP_001332309.1 12 - 13 2213 114
NM_001345492.1 NP_001332311.1 12 - 13 2188 114

View Full Product Details