Assay Design



Gene Transcripts

Gene Symbol
ALDH12A1
Entrez Gene ID
836373
Gene Name
aldehyde dehydrogenase 12A1
Gene Aliases
ARABIDOPSIS THALIANA DELTA1-PYRROLINE-5-CARBOXYLATE DEHYDROGENASE, AT5G62530, ATP5CDH, DELTA1-PYROLINE-5-CARBOXYLATE DEHYDROGENASE, DELTA1-PYRROLINE-5-CARBOXYLATE DEHYDROGENASE, K19B1.14, K19B1_14, P5CDH, aldehyde dehydrogenase 12A1
Chromosome Location
Chr.5: 25099003 - 25103312 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125647.4 NP_568955.1 6 - 7 665 89
GenBank mRNA AY065072.1 - 6 - 7 562 89
BT000746.1 - 6 - 7 595 89
BT010391.1 - 6 - 7 512 89

View Full Product Details