Assay Design



Gene Transcripts

Gene Symbol
RR19
Entrez Gene ID
841342
Gene Name
response regulator 19
Gene Aliases
ARR19, AT1G49190, F27J15.4, F27J15_4, response regulator 19
Chromosome Location
Chr.1: 18191267 - 18193782 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198250.1 NP_001185179.1 1 - 2 253 154
NM_001333372.1 NP_001319180.1 1 - 2 178 154

View Full Product Details