Assay Design
Gene Transcripts
- Gene Symbol
- NHD2
- Entrez Gene ID
-
841404
- Gene Name
- Na+/H+ antiporter 2
- Gene Aliases
-
AT1G49810, ATNHD2, F10F5.6, F10F5_6, Na+/H+ antiporter 2, SODIUM:HYDROGEN ANTIPORTER 2
- Chromosome Location
-
Chr.1: 18439945 - 18442237 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103868.2 |
NP_175403.1 |
2 - 3 |
289 |
111 |
View Full Product Details