Assay Design



Gene Transcripts

Gene Symbol
GSTF14
Entrez Gene ID
841409
Gene Name
glutathione S-transferase (class phi) 14
Gene Aliases
AT1G49860, ATGSTF14, F10F5.9, F10F5_9, glutathione S-transferase (class phi) 14
Chromosome Location
Chr.1: 18458526 - 18459969 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103873.5 NP_175408.1 1 - 2 338 77
GenBank mRNA AK176306.1 - 1 - 2 214 77
BT010408.1 - 1 - 2 154 77

View Full Product Details