Assay Design



Gene Transcripts

Gene Symbol
TOR
Entrez Gene ID
841427
Gene Name
target of rapamycin
Gene Aliases
AT1G50030, F2J10.19, F2J10_19, target of rapamycin
Chromosome Location
Chr.1: 18522248 - 18539995 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123987.1 NP_001117459.1 53 - 54 7146 157
NM_103891.4 NP_175425.2 53 - 54 7227 157
GenBank mRNA AF178967.1 - 53 - 54 7082 157

View Full Product Details