Assay Design



Gene Transcripts

Gene Symbol
FU
Entrez Gene ID
841446
Gene Name
kinase family with ARM repeat domain-containing protein
Gene Aliases
AT1G50240, FUSED
Chromosome Location
Chr.1: 18606511 - 18614500 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333427.1 NP_001319189.1 4 - 5 588 73
NM_001333428.1 NP_001321239.1 4 - 5 521 73

View Full Product Details