Assay Design
Gene Transcripts
- Gene Symbol
- VIIIA
- Entrez Gene ID
-
841458
- Gene Name
- P-loop containing nucleoside triphosphate hydrolases superfamily protein
- Gene Aliases
-
AT1G50360, ATVIIIA, F14I3.6, F14I3_6, MYOSIN VIII A
- Chromosome Location
-
Chr.1: 18649706 - 18657400 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103919.5 |
NP_175453.2 |
23 - 24 |
3260 |
122 |
View Full Product Details