Assay Design



Gene Transcripts

Gene Symbol
THFS
Entrez Gene ID
841470
Gene Name
10-formyltetrahydrofolate synthetase
Gene Aliases
10-FORMYLTETRAHYDROFOLATE SYNTHETASE, 10-formyltetrahydrofolate synthetase, AT1G50480, F11F12.17
Chromosome Location
Chr.1: 18701882 - 18704938 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103931.4 NP_564571.1 1 - 2 340 107
GenBank mRNA AF162279.1 - 1 - 2 161 107
AY054637.1 - 1 - 2 259 107
AY081549.1 - 1 - 2 158 107

View Full Product Details