Assay Design



Gene Transcripts

Gene Symbol
THFS
Entrez Gene ID
841470
Gene Name
10-formyltetrahydrofolate synthetase
Gene Aliases
10-FORMYLTETRAHYDROFOLATE SYNTHETASE, 10-formyltetrahydrofolate synthetase, AT1G50480, F11F12.17
Chromosome Location
Chr.1: 18701882 - 18704938 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103931.4 NP_564571.1 4 - 5 1817 65
GenBank mRNA AF162279.1 - 4 - 5 1638 65
AY054637.1 - 4 - 5 1736 65
AY081549.1 - 4 - 5 1635 65

View Full Product Details