Assay Design



Gene Transcripts

Gene Symbol
POLGAMMA2
Entrez Gene ID
841506
Gene Name
polymerase gamma 2
Gene Aliases
AT1G50840, F8A12.8, F8A12_8, PolIA, polymerase I A, polymerase gamma 2
Chromosome Location
Chr.1: 18839047 - 18844577 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333457.1 NP_001321058.1 3 - 4 1311 91
NM_103965.5 NP_175498.2 3 - 4 1266 91

View Full Product Details