Assay Design



Gene Transcripts

Gene Symbol
MIPS3
Entrez Gene ID
830881
Gene Name
myo-inositol-1-phosphate synthase 3
Gene Aliases
AT5G10170, ATMIPS3, MYO-INOSITOL-1-PHOSTPATE SYNTHASE 3, T31P16.160, T31P16_160, myo-inositol-1-phosphate synthase 3
Chromosome Location
Chr.5: 3187277 - 3190321 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121055.4 NP_196579.1 6 - 7 1143 73
GenBank mRNA AY065415.1 - 6 - 7 1073 73
AY096554.1 - 6 - 7 983 73

View Full Product Details