Assay Design
Gene Transcripts
- Gene Symbol
- DOT3
- Entrez Gene ID
-
830889
- Gene Name
- Phototropic-responsive NPH3 family protein
- Gene Aliases
-
AT5G10250, DEFECTIVELY ORGANIZED TRIBUTARIES 3, F18D22.20, F18D22_20
- Chromosome Location
-
Chr.5: 3216881 - 3219436 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121063.2 |
NP_196587.1 |
3 - 4 |
1163 |
119 |
View Full Product Details