Assay Design



Gene Transcripts

Gene Symbol
PAS2
Entrez Gene ID
830912
Gene Name
Protein-tyrosine phosphatase-like, PTPLA
Gene Aliases
AT5G10480, F12B17.170, F12B17_170, PASTICCINO 2, PEP, PEPINO
Chromosome Location
Chr.5: 3297804 - 3300165 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085097.1 NP_001078566.1 6 - 7 509 117
NM_001203348.1 NP_001190277.1 7 - 8 543 117
NM_121086.5 NP_196610.2 6 - 7 530 117
GenBank mRNA AJ506746.1 - 6 - 7 429 117
AY047708.1 - 6 - 7 496 117
AY065090.1 - 6 - 7 509 117
BT006269.1 - 6 - 7 413 117

View Full Product Details