Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161232.2 | NP_001154704.2 | 3 - 4 | 518 | 91 |
NM_001161233.1 | NP_001154705.1 | 3 - 4 | 477 | 91 | |
NM_001343149.1 | NP_001332043.1 | 3 - 4 | 480 | 91 | |
NM_001343150.1 | NP_001332042.1 | 3 - 4 | 480 | 91 | |
NM_001343151.1 | NP_001332040.1 | 3 - 4 | 480 | 91 | |
NM_001343152.1 | NP_001332041.1 | 3 - 4 | 477 | 91 | |
NM_001343153.1 | NP_001332045.1 | 3 - 4 | 477 | 91 | |
NM_001343154.1 | NP_001332044.1 | 3 - 4 | 477 | 91 | |
NM_121141.5 | NP_196664.4 | 3 - 4 | 518 | 91 |