Assay Design



Gene Transcripts

Gene Symbol
ALF4
Entrez Gene ID
830970
Gene Name
aberrant root formation protein
Gene Aliases
ABERRANT LATERAL ROOT FORMATION 4, AT5G11030, T5K6.20, T5K6_20
Chromosome Location
Chr.5: 3490788 - 3495173 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161233.1 NP_001154705.1 8 - 9 1255 63
NM_121141.5 NP_196664.4 8 - 9 1296 63

View Full Product Details