Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001343191.1 | NP_001331464.1 | 2 - 3 | 633 | 75 |
NM_121182.3 | NP_196705.1 | 2 - 3 | 407 | 75 | |
GenBank mRNA | AY735675.1 | - | 2 - 3 | 405 | 75 |
AY773894.1 | - | 1 - 2 | 320 | 75 |