Assay Design



Gene Transcripts

Gene Symbol
CPK7
Entrez Gene ID
831123
Gene Name
calmodulin-domain protein kinase 7
Gene Aliases
AT5G12480, CALMODULIN-DOMAIN PROTEIN KINASE 7, calmodulin-domain protein kinase 7
Chromosome Location
Chr.5: 4047513 - 4050643 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085107.1 NP_001078576.1 7 - 8 891 97
NM_121286.4 NP_568281.1 6 - 7 1368 97
GenBank mRNA AF361634.1 - 6 - 7 1259 97
U31836.1 - 6 - 7 1198 97

View Full Product Details