Assay Design



Gene Transcripts

Gene Symbol
WRKY60
Entrez Gene ID
817039
Gene Name
WRKY DNA-binding protein 60
Gene Aliases
AT2G25000, ATWRKY60, F27C12.8, F27C12_8, WRKY DNA-binding protein 60
Chromosome Location
Chr.2: 10629662 - 10631413 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335968.1 NP_001325298.1 3 - 4 903 74
NM_001335969.1 NP_001325299.1 3 - 4 565 74
NM_001335970.1 NP_001325297.1 3 - 4 586 74
NM_128058.4 NP_180072.1 4 - 5 699 74
GenBank mRNA AK227987.1 - 4 - 5 611 74
BT004611.1 - 4 - 5 549 74

View Full Product Details