Assay Design



Gene Transcripts

Gene Symbol
RLP21
Entrez Gene ID
817085
Gene Name
receptor like protein 21
Gene Aliases
AT2G25470, AtRLP21, F13B15.13, F13B15_13, receptor like protein 21
Chromosome Location
Chr.2: 10838294 - 10842016 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335998.1 NP_001325074.1 5 - 6 847 73
NM_001335999.1 NP_001325073.1 5 - 6 834 73
NM_128104.5 NP_180117.4 5 - 6 721 73

View Full Product Details