Assay Design



Gene Transcripts

Gene Symbol
TIP4;1
Entrez Gene ID
817123
Gene Name
tonoplast intrinsic protein 4;1
Gene Aliases
AT2G25810, F17H15.16, F17H15_16, tonoplast intrinsic protein 4;1
Chromosome Location
Chr.2: 11012499 - 11014171 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128141.6 NP_180152.1 2 - 3 530 67
GenBank mRNA AK226190.1 - 2 - 3 528 67
BT012635.1 - 2 - 3 371 67

View Full Product Details